CAS RN 20225-24-5
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
2-Ethylpentanoic Acid, TRC
CAS: 20225-24-5 Molekylær formel: C7 H14 O2 Molekylvægt (g/mol): 130.18 Synonym: (2RS)-2-Ethylpentanoic Acid,Valproic Acid Imp. B (EP) IUPAC navn: 2-ethylpentanoic acid SMIL: CCCC(CC)C(=O)O
2
(2RS)-2-Ethylpentanoic Acid, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.