CAS RN 192-65-4
IUPAC Navn:
hexacyclo[10.10.2.0²,⁷.0⁸,²⁴.0¹⁵,²³.0¹⁶,²¹]tetracosa-1(22),2,4,6,8,10,12,14,16,18,20,23-dodecaene
Synonymer:
dibenzopyrenes DIBENZO[a,e]PYRENE
Molekylvægt (g/mol):
302.38
Molekylformel:
C24H14
InChi Key:
KGHMWBNEMFNJFZ-UHFFFAOYSA-N
SMILES:
C1=CC=C2C(=C1)C=C1C3=CC=CC=C3C3=CC=CC4=CC=C2C1=C34
1
–
3
af
3
Resultater
1
Dibenzo[a,e]pyrene, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Dibenzo[a,e]pyrene, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.