CAS RN 1913269-12-1
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
KN-93 phosphate, MedChemExpress
MedChemExpress KN-93 phosphate is a novel membrane-permeant synthetic inhibitor of purified neuronal CaMK-II, with Ki of 370 nM.