CAS RN 191089-60-8
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
K-7174 dihydrochloride, MedChemExpress
MedChemExpress K-7174 dihydrochloride is a novel cell adhesion inhibitor; inhibits the expression of vascular cell adhesion molecule-1 (VCAM-1) induced by either IL-1β or TNF-α.