CAS RN 190386-37-9
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
N-Methylpyrrole-d4 (ring-d4), CDN
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
2
N-Methylpyrrole-d4 (ring-d4), TRC
CAS: 190386-37-9 Molekylær formel: C5 D4 H3 N Molekylvægt (g/mol): 85.14 Synonym: 1H-Pyrrole-2,3,4,5-d4, 1-methyl- (9CI),1-Methyl-1H-pyrrole-2,3,4,5-d4,1-Methylpyrrole-d4,N-Methylpyrrole-2,3,4,5-d4,N-Methylpyrrole-2,3,4,5-D4,N-Methylpyrrole-d4 (ring-d4) IUPAC navn: 2,3,4,5-tetradeuterio-1-methylpyrrole SMIL: [2H]c1c([2H])c([2H])n(C)c1[2H]