Få mere at vide
N-Methylpyrrole-d4 (ring-d4), TRC
Artikelnummer. 30483049
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30483049 100 mg
30465580 10 mg
30481061 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30483049 Leverandør Toronto Research Chemicals Leverandørnr. TRCM327016100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 190386-37-9
Molekylær formel C5 D4 H3 N
Molekylvægt (g/mol) 85.14
Synonym 1H-Pyrrole-2,3,4,5-d4, 1-methyl- (9CI), 1-Methyl-1H-pyrrole-2,3,4,5-d4, 1-Methylpyrrole-d4, N-Methylpyrrole-2,3,4,5-d4, N-Methylpyrrole-2,3,4,5-D4, N-Methylpyrrole-d4 (ring-d4)
IUPAC navn 2,3,4,5-tetradeuterio-1-methylpyrrole
SMIL [2H]c1c([2H])c([2H])n(C)c1[2H]
Tekniske data
Kemisk navn eller materiale N-Methylpyrrole-d4 (ring-d4)
Mængde 100 mg
InChI formel InChI=1S/C5H7N/c1-6-4-2-3-5-6/h2-5H,1H3/i2D,3D,4D,5D
Formel vægt 85.083