Få mere at vide
N-Methylpyrrole-d4 (ring-d4), CDN
Artikelnummer. 30317489
Skift visning
Mængde:
0.5 g
1 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30317489 1 g -
30309081 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30317489 Leverandør CDN Leverandørnr. CDND58321G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Methylpyrrole-d4 (ring-d4)
CAS 190386-37-9
Molekylær formel C5 D4 H3 N
Mængde 1 g
Synonym 1 H-Pyrrole-2,3,4,5-d4, 1-methyl- (9 CI), 1-Methyl-1 H-pyrrole-2,3,4,5-d4, 1-Methylpyrrole-d4, N-Methylpyrrole-2,3,4,5-d4, N-Methylpyrrole-2,3,4,5-D4, N-Methylpyrrole-d4 (ring-d4)
InChI formel InChI=1 S/C5H7N/c1-6-4-2-3-5-6/h2-5 H,1H3/i2D,3 D,4 D,5 D
SMIL [2 H]c1c([2 H])c([2 H])n(C)c1[2 H]
IUPAC navn 2,3,4,5-tetradeuterio-1-methylpyrrole
Molekylvægt (g/mol) 85.14
Formel vægt 85.083 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity