CAS RN 188396-77-2
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Paliroden, MedChemExpress
MedChemExpress Paliroden is an orally bioactive neurotrophic, non-peptidic compound that activates synthesis of endogenous neurotrophines, used for treatment of Alzheimer's Disease and Parkinson's.