CAS RN 1883545-52-5
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
Tocris Bioscience™ MM 102
WDR5/MLL interaction inhibitor
2
MM-102 TFA, MedChemExpress
MedChemExpress MM-102 TFA (HMTase Inhibitor IX TFA) is a potent WDR5/MLL interaction inhibitor, achieves IC50 = 2.4 nM with an estimated Ki 200 times more potent than the ARA peptide.
3