CAS RN 187034-31-7
IUPAC Navn:
N-[1-({1-[(2-aminoethyl)carbamoyl]ethyl}carbamoyl)-2,2-dimethylpropyl]-N'-hydroxy-2-(2-methylpropyl)butanediamide
Synonymer:
N-[1-({1-[(2-aminoethyl)carbamoyl]ethyl}carbamoyl)-2,2-dimethylpropyl]-N'-hydroxy-2-(2-methylpropyl)succinamide
Molekylvægt (g/mol):
415.54
Molekylformel:
C19H37N5O5
InChi Key:
LMIQCBIEAHJAMZ-UHFFFAOYNA-N
SMILES:
CC(C)CC(CC(=O)NO)C(=O)NC(C(=O)NC(C)C(=O)NCCN)C(C)(C)C
1
–
2
af
2
Resultater
1
TAPI-2, MedChemExpress
MedChemExpress TAPI-2 (TNF Protease Inhibitor 2) is a broad-spectrum inhibitor of matrix metalloprotease (MMP), tumour necrosis factorα-converting enzyme (TACE) and a disintegrin and metalloproteinase (ADAM), with an IC50 of 20 μM for MMP. TAPI-2 blocks the entry of infectious SARS-CoV.