missing translation for 'onlineSavingsMsg'
Få mere at vide

TAPI-2, MedChemExpress

Artikelnummer. 16495587
Encompass_Preferred
Change view
Click to view available options
Mængde:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
16495587 5 mg
16470549 1 mg
16452848 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16495587 Supplier Medchem Express Supplier No. HY100211

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress TAPI-2 (TNF Protease Inhibitor 2) is a broad-spectrum inhibitor of matrix metalloprotease (MMP), tumour necrosis factorα-converting enzyme (TACE) and a disintegrin and metalloproteinase (ADAM), with an IC50 of 20 μM for MMP. TAPI-2 blocks the entry of infectious SARS-CoV.

TAPI-2 (TNF Protease Inhibitor 2) is a broad-spectrum inhibitor of matrix metalloprotease (MMP), tumour necrosis factorα-converting enzyme (TACE) and a disintegrin and metalloproteinase (ADAM), with an IC50 of 20 μM for MMP. TAPI-2 blocks the entry of infectious SARS-CoV.

Specifications

Kemisk navn eller materiale TAPI-2
Fysisk form Solid
CAS 187034-31-7
Farve White
Mængde 5 mg
Renhedsgrad noter Research
Molekylær formel C19H37N5O5
Til brug med (applikation) Cancer-Kinase/protease
Synonym TNF Protease Inhibitor 2
Opløselighedsinformation H2O : 100 mg/mL (240.66 mM; Need ultrasonic) ∣DMSO : ≥ 22 mg/mL (52.94 mM)
SMIL CC(C)(C)[C@@H](C(N[C@H](C(NCCN)=O)C)=O)NC(C(CC(NO)=O)CC(C)C)=O
Molekylvægt (g/mol) 415.53
Formel vægt 415.53
Procent renhed 95.0%
Grad Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.