CAS RN 1780259-94-0
IUPAC Navn:
2-(2-{4-[(3-phenoxyphenyl)methyl]piperazin-1-yl}ethoxy)ethan-1-ol dihydrochloride
Synonymer:
2-(2-{4-[(3-phenoxyphenyl)methyl]piperazin-1-yl}ethoxy)ethanol dihydrochloride
Molekylvægt (g/mol):
429.38
Molekylformel:
C21H30Cl2N2O3
InChi Key:
MPACCEKWFGWZHS-UHFFFAOYSA-N
SMILES:
Cl.Cl.OCCOCCN1CCN(CC2=CC(OC3=CC=CC=C3)=CC=C2)CC1
1
–
1
af
1
Resultater
1
ZK756326 dihydrochloride, MedChemExpress
MedChemExpress ZK756326 dihydrochloride is a nonpeptide chemokine receptor agonist for the CC chemokine receptor CCR8.