CAS RN 177034-57-0
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
4-[(2-Isopropoxyethoxy)methyl]phenol, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
4-[(2-Isopropoxyethoxy)methyl]phenol, TRC
CAS: 177034-57-0 Molekylær formel: C12 H18 O3 Molekylvægt (g/mol): 210.27 Synonym: 4-[(2-Isopropoxyethoxy)methyl]phenol,Bisoprolol Fumarate Imp. M (Pharmeuropa) IUPAC navn: 4-(2-propan-2-yloxyethoxymethyl)phenol SMIL: CC(C)OCCOCc1ccc(O)cc1