CAS RN 17692-31-8
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
Dropropizine, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
rac Dropropizine, TRC
CAS: 17692-31-8 Molekylær formel: C13 H20 N2 O2 Molekylvægt (g/mol): 236.31 Synonym: Dropropizine IUPAC navn: 3-(4-phenylpiperazin-1-yl)propane-1,2-diol SMIL: OCC(O)CN1CCN(CC1)c2ccccc2