missing translation for 'onlineSavingsMsg'
Få mere at vide

Dropropizine, Mikromol™

Artikelnummer. 17922886
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17922886 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 17922886 Leverandør Mikromol Leverandørnr. MM1017.000250

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Dropropizine
TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Dropropizine
CAS 17692-31-8
Analyt- eller komponentnavne Dropropizine
Forsendelsestilstand Room Temperature
Molekylær formel C13 H20 N2 O2
Certificeringer/compliance ISO 17034
Synonym Dropropizine
InChI formel InChI=1S/C13H20N2O2/c16-11-13(17)10-14-6-8-15(9-7-14)12-4-2-1-3-5-12/h1-5,13,16-17H,6-11H2
SMIL OCC(O)CN1CCN(CC1)c2ccccc2
IUPAC navn 3-(4-phenylpiperazin-1-yl)propane-1,2-diol
Molekylvægt (g/mol) 236.31
Mængde 250 mg
Formel vægt 236.1525 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.