CAS RN 17685-05-1
IUPAC Navn:
(2S,3S,4S,5R,6S)-3,4,5-trihydroxy-6-phenoxyoxane-2-carboxylate
Synonymer:
(2S,3S,4S,5R,6S)-3,4,5-trihydroxy-6-phenoxyoxane-2-carboxylate
Molekylvægt (g/mol):
269.23
Molekylformel:
C12H13O7
InChi Key:
WVHAUDNUGBNUDZ-GOVZDWNOSA-M
SMILES:
O[C@@H]1[C@@H](O)[C@H](OC2=CC=CC=C2)O[C@@H]([C@H]1O)C([O-])=O
1
–
3
af
3
Resultater
1
Phenyl b-D-Glucuronide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Phenyl b-D-Glucuronide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Phenyl beta-D-Glucuronide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.