missing translation for 'onlineSavingsMsg'
Få mere at vide

Phenyl b-D-Glucuronide, TRC

Artikelnummer. 30412260
Change view
Click to view available options
Mængde:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30412260 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30412260 Supplier Toronto Research Chemicals Supplier No. TRCP32650025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Phenyl beta-D-Glucuronide
CAS 17685-05-1
Molekylær formel C12 H14 O7
Mængde 25 mg
Synonym Phenol-Glucuronide
InChI formel InChI=1S/C12H14O7/c13-7-8(14)10(11(16)17)19-12(9(7)15)18-6-4-2-1-3-5-6/h1-5,7-10,12-15H,(H,16,17)/t7-,8-,9+,10-,12+/m0/s1
SMIL O[C@@H]1[C@@H](O)[C@H](Oc2ccccc2)O[C@@H]([C@H]1O)C(=O)O
IUPAC navn (2S,3S,4S,5R,6S)-3,4,5-trihydroxy-6-phenoxyoxane-2-carboxylic acid
Molekylvægt (g/mol) 270.24
Formel vægt 270.074
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.