CAS RN 175278-21-4
IUPAC Navn:
2-(trifluoromethoxy)benzene-1,4-dicarboxylic acid
Synonymer:
2-(trifluoromethoxy)benzene-1,4-dicarboxylic acid
Molekylvægt (g/mol):
250.13
Molekylformel:
C9H5F3O5
InChi Key:
VVTIYBRREAGBBA-UHFFFAOYSA-N
SMILES:
OC(=O)C1=CC(OC(F)(F)F)=C(C=C1)C(O)=O
1
–
3
af
3
Resultater
1
2-(Trifluoromethoxy)terephthalic Acid (>85%), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2-(Trifluoromethoxy)terephthalic Acid (>85%), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
2-(Trifluoromethoxy)terephthalic Acid (>85%), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.