CAS RN 1731-94-8
IUPAC Navn:
methyl nonadecanoate
Synonymer:
methyl nonadecanoate
Molekylvægt (g/mol):
312.54
Molekylformel:
C20H40O2
InChi Key:
BDXAHSJUDUZLDU-UHFFFAOYSA-N
SMILES:
CCCCCCCCCCCCCCCCCCC(=O)OC
1
–
2
af
2
Resultater
1
Methyl Nonadecanoate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.