CAS RN 172939-67-2
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
3-O-Benzoyl-N-acetyl-α-D-galactosaminyl-1-O-N-(Fmoc)serine Phenacylester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
3-O-Benzoyl-N-acetyl-α-D-galactosaminyl-1-O-N-(Fmoc)serine Phenacylester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
3-O-Benzoyl-N-acetyl-α-D-galactosaminyl-1-O-N-(Fmoc)serine Phenacylester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.