CAS RN 1676893-24-5
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
NSC781406, MedChemExpress
MedChemExpress NSC781406 is a highly potent PI3K and mTOR inhibitor with an IC50 of 2 nM for PI3Kα.