CAS RN 16416-29-8
IUPAC Navn:
decane
Synonymer:
decane n-dekan decane (substance) dekan ccris 653 N-decane hsdb 63
Molekylvægt (g/mol):
142.29
Molekylformel:
C10H22
InChi Key:
DIOQZVSQGTUSAI-UHFFFAOYSA-N
SMILES:
CCCCCCCCCC
1
–
4
af
4
Resultater
1
N-Decane-d22, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
N-Decane-d22, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
N-Decane-d22, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
n-Decane-d22, CDN
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.