Få mere at vide
N-Decane-d22, TRC
Artikelnummer. 30352255
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30352255 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30352255 Leverandør Toronto Research Chemicals Leverandørnr. TRCD22634550MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale n-Decane-d22
CAS 16416-29-8
Molekylær formel C10 D22
Mængde 50 mg
Synonym Decane-1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-d22 (ACI), Decane-d22 (8CI, 9CI), Perdeutero-n-decane, Perdeuterodecane, Decane-D22
InChI formel InChI=1S/C10H22/c1-3-5-7-9-10-8-6-4-2/h3-10H2,1-2H3/i1D3,2D3,3D2,4D2,5D2,6D2,7D2,8D2,9D2,10D2
SMIL [2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H]
IUPAC navn 1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-docosadeuteriodecane
Molekylvægt (g/mol) 164.42
Formel vægt 164.31