CAS RN 1622848-92-3
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
GSK2982772, MedChemExpress
MedChemExpress GSK2982772 is a potent, orally active and ATP competitive RIP1 kinase inhibitor with IC50 values of 16 nM and 20 nM for human and monkey RIP1, respectively.