CAS RN 1620401-83-3
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
UNC0379 TFA, MedChemExpress
MedChemExpress UNC0379 TFA is a selective, substrate-competitive inhibitor of lysine methyltransferase SETD8 (KMT5A) with an IC50 of 7.3 μM, KD value of 18.3 μM. UNC0379 TFA can be used in the research of inflammation and cancers, such as pulmonary fibrosis, ovarian cancer, neuroblastoma.