CAS RN 1615713-87-5
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
DprE1-IN-2, MedChemExpress
MedChemExpress DprE1-IN-2 (compound 18) is a potent DprE1 inhibitor with an IC50 of 28 nM. DprE1-IN-2 has antituberculosis effect.