CAS RN 1610537-15-9
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Seviteronel, MedChemExpress
MedChemExpress Seviteronel (VT-464) is a potent CYP17 lyase inhibitor(h-Lyase IC50=69 nM) that demonstrated both exceptional in vitro lyase/hydroxylase selectivity (∼10-fold) and oral activity in a hamster model of androgen biosynthesis inhibition.