CAS RN 1599432-08-2
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
NIBR189, MedChemExpress
MedChemExpress NIBR189 is a small molecule antagonist of the Epstein-Barr virus-induced gene 2 (EBI2; GPR183) receptor with IC50 of 16 nM(Binding) and 11 nM (Functional).