CAS RN 15917-65-4
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
N-Desmethyl Tamoxifen Hydrochloride, TRC
CAS: 15917-65-4 Molekylær formel: C25 H27 N O . Cl H Molekylvægt (g/mol): 393.95 Synonym: 2-[4-[(Z)-1,2-Diphenylbut-1-enyl]phenoxy]-N-methylethanamine Hydrochloride,Tamoxifen Citrate Imp. F (EP) as Hydrochloride IUPAC navn: 2-[4-[(Z)-1,2-diphenylbut-1-enyl]phenoxy]-N-methylethanamine;hydrochloride SMIL: Cl.CC\C(=C(/c1ccccc1)\c2ccc(OCCNC)cc2)\c3ccccc3
2
Tamoxifen Citrate EP Impurity F (as Hydrochloride), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.