Få mere at vide
N-Desmethyl Tamoxifen Hydrochloride, TRC
Artikelnummer. 30136602
Skift visning
Mængde:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30136602 25 mg
30005695 10 mg
30005863 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30136602 Leverandør Toronto Research Chemicals Leverandørnr. TRCD29390025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 15917-65-4
Molekylær formel C25 H27 N O . Cl H
Molekylvægt (g/mol) 393.95
Synonym 2-[4-[(Z)-1,2-Diphenylbut-1-enyl]phenoxy]-N-methylethanamine Hydrochloride, Tamoxifen Citrate Imp. F (EP) as Hydrochloride
IUPAC navn 2-[4-[(Z)-1,2-diphenylbut-1-enyl]phenoxy]-N-methylethanamine;hydrochloride
SMIL Cl.CC\C(=C(/c1ccccc1)\c2ccc(OCCNC)cc2)\c3ccccc3
Tekniske data
Kemisk navn eller materiale 2-[4-[(Z)-1,2-Diphenylbut-1-enyl]phenoxy]-N-methylethanamine Hydrochloride
Renhedsgrad noter HPLC
Mængde 25 mg
InChI formel InChI=1S/C25H27NO.ClH/c1-3-24(20-10-6-4-7-11-20)25(21-12-8-5-9-13-21)22-14-16-23(17-15-22)27-19-18-26-2;/h4-17,26H,3,18-19H2,1-2H3;1H/b25-24-;
Formel vægt 393.1859
Procent renhed >95%