Få mere at vide
Tamoxifen Citrate EP Impurity F (as Hydrochloride), Mikromol™
Beskrivelse
Tamoxifen Citrate EP Impurity F (as Hydrochloride), Mikromol
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 2-[4-[(Z)-1,2-Diphenylbut-1-enyl]phenoxy]-N-methylethanamine Hydrochloride |
| CAS | 15917-65-4 |
| Analyt- eller komponentnavne | 2-[4-[(Z)-1,2-Diphenylbut-1-enyl]phenoxy]-N-methylethanamine Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C25 H27 N O . Cl H |
| Certificeringer/compliance | ISO 17025 |
| Synonym | 2-[4-[(Z)-1,2-Diphenylbut-1-enyl]phenoxy]-N-methylethanamine Hydrochloride, Tamoxifen Citrate Imp. F (EP) as Hydrochloride, Tamoxifen Citrate Impurity F as Hydrochloride, Tamoxifen Impurity F as Hydrochloride |
| InChI formel | InChI=1S/C25H27NO.ClH/c1-3-24(20-10-6-4-7-11-20)25(21-12-8-5-9-13-21)22-14-16-23(17-15-22)27-19-18-26-2;/h4-17,26H,3,18-19H2,1-2H3;1H/b25-24-; |
| Løsningstype | Neat |
| SMIL | Cl.CC\C(=C(/c1ccccc1)\c2ccc(OCCNC)cc2)\c3ccccc3 |
| Vis mere |