CAS RN 156453-53-1
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
4
af
4
Resultater
1
(aR)-a-[2-(1-Naphthalenyloxy)ethyl]benzenemethanol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(aR)-a-[2-(1-Naphthalenyloxy)ethyl]benzenemethanol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(1R)-3-(1-Naphthyloxy)-1-phenylpropan-1-ol, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
4
(alphaR)-alpha-[2-(1-Naphthalenyloxy)ethyl]benzenemethanol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.