Få mere at vide
(1R)-3-(1-Naphthyloxy)-1-phenylpropan-1-ol, Mikromol™
Artikelnummer. 30050606
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30050606 25 mg
30050077 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30050606 Leverandør Mikromol Leverandørnr. MM3523.010025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(1R)-3-(1-Naphthyloxy)-1-phenylpropan-1-ol
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (1R)-3-(1-Naphthyloxy)-1-phenylpropan-1-ol
CAS 156453-53-1
Analyt- eller komponentnavne (1R)-3-(1-Naphthyloxy)-1-phenylpropan-1-ol
Forsendelsestilstand Room Temperature
Molekylær formel C19 H18 O2
Certificeringer/compliance ISO 17025
Synonym (αR)-α-[2-(1-naphthalenyloxy)ethyl]benzenemethanol
InChI formel InChI=1S/C19H18O2/c20-18(16-8-2-1-3-9-16)13-14-21-19-12-6-10-15-7-4-5-11-17(15)19/h1-12,18,20H,13-14H2/t18-/m1/s1
SMIL O[C@H](CCOc1cccc2ccccc12)c3ccccc3
IUPAC navn (1R)-3-naphthalen-1-yloxy-1-phenylpropan-1-ol
Molekylvægt (g/mol) 278.34
Mængde 25 mg
Formel vægt 278.1307 g/mol