CAS RN 155593-75-2
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
Perphenazine-d4 (phenothiazine-1,3,7,9-d4), TRC
CAS: 155593-75-2 Molekylær formel: C21 D4 H22 Cl N3 O S Molekylvægt (g/mol): 407.99 Synonym: 1-Piperazineethanol, 4-[3-(2-chloro-10H-phenothiazin-10-yl-1,3,7,9-d4)propyl]- (9CI) IUPAC navn: 2-[4-[3-(2-chloro-1,3,7,9-tetradeuteriophenothiazin-10-yl)propyl]piperazin-1-yl]ethanol SMIL: [2H]c1cc([2H])c2N(CCCN3CCN(CCO)CC3)c4c([2H])c(Cl)c([2H])cc4Sc2c1
2
Perphenazine-d4 (phenothiazine-1,3,7,9-d4), CDN
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.