Få mere at vide
Perphenazine-d4 (phenothiazine-1,3,7,9-d4), CDN
Artikelnummer. 30314581
Skift visning
Mængde:
0.01 g
0.05 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30314581 0.05 g
30309055 0.01 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30314581 Leverandør CDN Leverandørnr. CDND27500.05G
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Perphenazine-d4 (phenothiazine-1,3,7,9-d4)
CAS 155593-75-2
Molekylær formel C21 D4 H22 Cl N3 O S
Mængde 0.05 g
Synonym 1-Piperazineethanol, 4-[3-(2-chloro-10 H-phenothiazin-10-yl-1,3,7,9-d4)propyl]- (9 CI)
InChI formel InChI=1 S/C21H26ClN3OS/c22-17-6-7-21-19(16-17)25(18-4-1-2-5-20(18)27-21)9-3-8-23-10-12-24(13-11-23)14-15-26/h1-2,4-7,16,26 H,3,8-15H2/i2D,4 D,6 D,16 D
SMIL [2 H]c1cc([2 H])c2N(CCCN3CCN(CCO)CC3)c4c([2 H])c(Cl)c([2 H])cc4Sc2c1
IUPAC navn 2-[4-[3-(2-chloro-1,3,7,9-tetradeuteriophenothiazin-10-yl)propyl]piperazin-1-yl]ethanol
Molekylvægt (g/mol) 407.99
Formel vægt 407.1736 g/mol
Procent renhed 98 atom % D, min 97% Chemical Purity