Få mere at vide
Perphenazine-d4 (phenothiazine-1,3,7,9-d4), TRC
Artikelnummer. 30425286
Skift visning
Mængde:
0.5 mg
1 mg
5 mg
Pakningsstørrelse:
0.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30425286 5 mg -
30418222 1 mg -
30409911 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30425286 Leverandør Toronto Research Chemicals Leverandørnr. TRCP2911015MG
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 155593-75-2
Molekylær formel C21 D4 H22 Cl N3 O S
Molekylvægt (g/mol) 407.99
Synonym 1-Piperazineethanol, 4-[3-(2-chloro-10H-phenothiazin-10-yl-1,3,7,9-d4)propyl]- (9CI)
IUPAC navn 2-[4-[3-(2-chloro-1,3,7,9-tetradeuteriophenothiazin-10-yl)propyl]piperazin-1-yl]ethanol
SMIL [2H]c1cc([2H])c2N(CCCN3CCN(CCO)CC3)c4c([2H])c(Cl)c([2H])cc4Sc2c1
Tekniske data
Kemisk navn eller materiale Perphenazine-d4 (phenothiazine-1,3,7,9-d4)
Mængde 5 mg
InChI formel InChI=1S/C21H26ClN3OS/c22-17-6-7-21-19(16-17)25(18-4-1-2-5-20(18)27-21)9-3-8-23-10-12-24(13-11-23)14-15-26/h1-2,4-7,16,26H,3,8-15H2/i2D,4D,6D,16D
Formel vægt 407.1736