CAS RN 1554458-53-5
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
BAY1217389, MedChemExpress
MedChemExpress BAY 1217389 is a potent, and selective inhibitor of the monopolar spindle 1 (MPS1) kinase with an IC50 value less than 10 nM.