CAS RN 1553-56-6
IUPAC Navn:
(4R)-4-[(1R,3aS,3bR,5aR,9aS,9bS,11aR)-9a,11a-dimethyl-7-oxo-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanoic acid
Synonymer:
3-ketolithocholic acid 3-oxocholan-24-oic acid
Molekylvægt (g/mol):
374.57
Molekylformel:
C24H38O3
InChi Key:
KIQFUORWRVZTHT-OPTMKGCMSA-N
SMILES:
C[C@H](CCC(O)=O)[C@H]1CC[C@H]2[C@@H]3CC[C@@H]4CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C
1
–
2
af
2
Resultater
1
3-Oxo-5b-cholanoic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
3-Oxo-5beta-cholanoic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.