missing translation for 'onlineSavingsMsg'
Learn More

3-Oxo-5b-cholanoic Acid, TRC

Product Code. 30426124
Change view
Click to view available options
Mængde:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30426124 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30426124 Supplier Toronto Research Chemicals Supplier No. TRCO8484905G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 3-Oxo-5beta-cholanoic Acid
CAS 1553-56-6
Renhedsgrad noter HPLC
Molekylær formel C24 H38 O3
Mængde 5 g
Synonym 3-Oxo-5β-cholan-24-oic Acid, 3-Oxo- 5β-cholanic Acid, 3-Ketolithocholic Acid, 3-Oxo-5β-cholan-24-oic Acid, 3-Oxo-5β-cholanic acid, 3-Oxo-5β-cholanoic Acid, 3-Oxocholanic Acid, 3-Oxolithocholic Acid, 3-keto-5β-Cholan-24-oic Acid, 3-keto-5β-Cholanic Acid, 5β-Cholanic Acid-3-one, Dehydrolithocholic Acid, Dehydro-lithocholic Acid
InChI formel InChI=1S/C24H38O3/c1-15(4-9-22(26)27)19-7-8-20-18-6-5-16-14-17(25)10-12-23(16,2)21(18)11-13-24(19,20)3/h15-16,18-21H,4-14H2,1-3H3,(H,26,27)/t15-,16-,18+,19-,20+,21+,23+,24-/m1/s1
SMIL C[C@H](CCC(=O)O)[C@H]1CC[C@H]2[C@@H]3CC[C@@H]4CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C
IUPAC navn (4R)-4-[(5R,8R,9S,10S,13R,14S,17R)-10,13-dimethyl-3-oxo-1,2,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]pentanoic acid
Molekylvægt (g/mol) 374.56
Formel vægt 374.2821
Procent renhed >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.