CAS RN 1527-97-5
IUPAC Navn:
(2E)-1-(2,4-dinitrophenyl)-2-hexylidenehydrazine
Synonymer:
(2E)-1-(2,4-dinitrophenyl)-2-hexylidenehydrazine
Molekylvægt (g/mol):
280.28
Molekylformel:
C12H16N4O4
InChi Key:
SJUYYAXVFFVQNX-MDWZMJQESA-N
SMILES:
CCCCC\C=N\NC1=CC=C(C=C1[N+]([O-])=O)[N+]([O-])=O
1
–
2
af
2
Resultater
1
Hexanal-2,4-dinitrophenylhydrazone, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
2
Hexanal-DNPH 1000 μg/mL in Methanol, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality