CAS RN 150821-03-7
IUPAC Navn:
bis(N-[4-oxo-2-(2H-1,2,3,4-tetrazol-5-yl)-4H-chromen-8-yl]-4-(4-phenylbutoxy)benzamide) hydrate
Synonymer:
bis(pranlukast) hydrate
Molekylvægt (g/mol):
981.04
Molekylformel:
C54H48N10O9
InChi Key:
MSXTUBJFNBZPGC-UHFFFAOYSA-N
SMILES:
O.O=C(NC1=C2OC(=CC(=O)C2=CC=C1)C1=NNN=N1)C1=CC=C(OCCCCC2=CC=CC=C2)C=C1.O=C(NC1=C2OC(=CC(=O)C2=CC=C1)C1=NNN=N1)C1=CC=C(OCCCCC2=CC=CC=C2)C=C1
1
–
2
af
2
Resultater
1
Thermo Scientific Chemicals Pranlukast hemihydrat
CAS: 150821-03-7 Molekylær formel: C54H48N10O9 Molekylvægt (g/mol): 981.04 InChI nøgle: MSXTUBJFNBZPGC-UHFFFAOYSA-N IUPAC navn: bis(N-[4-oxo-2-(2H-1,2,3,4-tetrazol-5-yl)-4H-chromen-8-yl]-4-(4-phenylbutoxy)benzamid)hydrat SMIL: O.O=C(NC1=C2OC(=CC(=O)C2=CC=C1)C1=NNN=N1)C1=CC=C(OCCCCC2=CC=CC=C2)C=C1.O=C(NC1=C2OC(=CC(=O)C2=CC=C1)C1=NNN=N1)C1=CC=C(OCCCCC2=CC=CC=C2)C=C1
2
Pranlukast hemihydrate, MedChemExpress
MedChemExpress Pranlukast hemihydrate is a highly potent, selective and competitive antagonist of peptide leukotrienes. Pranlukast inhibits [3H]LTE4, [3H]LTD4, and [3H]LTC4 bindings to lung membranes with Kis of 0.63±0.11, 0.99±0.19, and 5640±680 nM, respectively.