CAS RN 148891-53-6
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
6-(5-Chloropyridin-2-yl)-6,7-dihydro-5H-pyrrolo[3,4-b]pyrazin-5-one, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
RP 48497 (Eszopiclone Impurity C), TRC
CAS: 148891-53-6 Molekylær formel: C11 H7 Cl N4 O Molekylvægt (g/mol): 246.65 Synonym: 6-(5-Chloropyridin-2-yl)-6,7-dihydro-5H-pyrrolo[3,4-b]pyrazin-5-one,Zopiclone Imp. C (EP) IUPAC navn: 6-(5-chloropyridin-2-yl)-7H-pyrrolo[3,4-b]pyrazin-5-one SMIL: Clc1ccc(nc1)N2Cc3nccnc3C2=O