CAS RN 146651-75-4
IUPAC Navn:
tert-butyl N-(2-aminophenyl)carbamate
Synonymer:
tert-butyl N-(2-aminophenyl)carbamate
Molekylvægt (g/mol):
208.26
Molekylformel:
C11H16N2O2
InChi Key:
KCZFBLNQOSFGSH-UHFFFAOYSA-N
SMILES:
CC(C)(C)OC(=O)NC1=CC=CC=C1N
1
–
1
af
1
Resultater
1
N-Boc-1,2-phenyldiamine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.