CAS RN 14618-78-1
IUPAC Navn:
4,4-dimethoxybutanenitrile
Synonymer:
4,4-dimethoxybutanenitrile
Molekylvægt (g/mol):
129.16
Molekylformel:
C6H11NO2
InChi Key:
XJHNCGVHPLDMRK-UHFFFAOYSA-N
SMILES:
COC(CCC#N)OC
1
–
1
af
1
Resultater
1
4,4-Dimethoxybutyronitrile, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.