CAS RN 1459687-96-7
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
GJ103 sodium salt, MedChemExpress
MedChemExpress GJ103 sodium salt is an active analog of the read-through compound GJ072.