CAS RN 1454288-88-0
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Rovazolac, MedChemExpress
MedChemExpress Rovazolac is a liver x receptor (LXR) modulator extracted from patent WO2013130892A1.