CAS RN 1446194-56-4
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
4
af
4
Resultater
1
N-[(1S)-2-[(3-Methylbutyl)amino]-2-oxo-1-(phenylmethyl)ethyl]-2-pyrazinecarboxamide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
N-[(1S)-2-[(3-Methylbutyl)amino]-2-oxo-1-(phenylmethyl)ethyl]-2-pyrazinecarboxamide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
N-[(1S)-2-[(3-Methylbutyl)amino]-2-oxo-1-(phenylmethyl)ethyl]-2-pyrazinecarboxamide, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
N-[(1S)-2-[(3-Methylbutyl)amino]-2-oxo-1-(phenylmethyl)ethyl]-2-pyrazinecarboxamide, Mikromol
Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.