CAS RN 1440251-53-5
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
Tocris Bioscience™ ML 315 hydrochloride
Low molecular weight, organic compound that may help regulate biological process.
2
Tocris Bioscience™ ML 315 hydrochloride
Low molecular weight, organic compound that may help regulate biological process.
3
Tocris Bioscience™ ML 315 hydrochloride
Low molecular weight, organic compound that may help regulate biological process.