Få mere at vide
Tocris Bioscience™ ML 315 hydrochloride
GREENER_CHOICE
Low molecular weight, organic compound that may help regulate biological process.
Brand:  Tocris Bioscience™ 5700/50
Encompass
Produktkode 15727066
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter »
Denne vare kan ikke returneres. Se returpolitik
Beskrivelse
Beskrivelse
ML 315 hydrochloride is a inhibitor of cdc2-like kinases (Clk) and DYRK kinases (IC50 values are 68, 68 and 231 nM for Clk1, Clk4 and Clk2, respectively, and 282 and 1156 nM for Dyrk1A and Dyrk1B). Shows 20-fold selectivity for Clk4 over Dyrk1A. Stable in both mouse and human plasma. Moderately cell permeable.
Tekniske data
Tekniske data
Inhibitor of Clk and DYRK Kinases
1440251-53-5
410.68g/mol
Cdc2-like Kinase Inhibitors
Cdc2-like Kinase
C18H13Cl2N3O2.HCl
Store at -20°C
50 mg
Produktforslag
Produktforslag
Videoer
SDS
Dokumenter
Dokumenter
Certifikater
Kampagner
Kampagner
Produkttitel
Vælg et problem
For Research Use Only