Få mere at vide
Tocris Bioscience™ ML 315 hydrochloride
Low molecular weight, organic compound that may help regulate biological process.
Brand: Tocris Bioscience™ 5700/10
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
ML 315 hydrochloride is a inhibitor of cdc2-like kinases (Clk) and DYRK kinases (IC50 values are 68, 68 and 231 nM for Clk1, Clk4 and Clk2, respectively, and 282 and 1156 nM for Dyrk1A and Dyrk1B). Shows 20-fold selectivity for Clk4 over Dyrk1A. Stable in both mouse and human plasma. Moderately cell permeable.Tekniske data
| Inhibitor of Clk and DYRK Kinases | |
| 1440251-53-5 | |
| 410.68g/mol | |
| Cdc2-like Kinase Inhibitors |
| Cdc2-like Kinase | |
| C18H13Cl2N3O2.HCl | |
| Store at -20°C | |
| 10 mg |
Produkttitel
Ser du en mulighed for forbedring?
For Research Use Only